missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ GeneJuice™, Transfection Reagent

Catalog No. p-7078295
Change view
Click to view available options
Quantity:
0.3 mL
1 mL
5 mL
10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
709673 1 mL
709674 10 mL
709675 0.3 mL
709676 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 709673 Supplier MilliporeSigma™ Supplier No. 709673
Only null left

Enables efficient DNA transfer in both stable transient transfections of eukaryotic cells.

EMD Millipore Novagen™ GeneJuice™, Transfection Reagent is non-lipid based chemical transfection reagent.

  • Efficient DNA transfer for both stable transient transfections
  • Minimal cellular toxicity
  • Compatibility with both serum-containing serum-free media
  • Simple protocol
  • Suitable for high-throughput transfection in multi-well plate format
  • CAS: Ethanol: 64-17-5
  • Packaging: Glass Bottle

Specifications

Content And Storage 2°C to 8°C
Quantity 1 mL