missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ CytoBuster™ Protein Extraction Reagent
Formulation of detergents optimized for efficient extraction of soluble proteins from mammalian and insect cells
Supplier: MilliporeSigma™ MIL710093
Description
- Gentle, non-ionic composition of reagent enables isolation of functionally active endogenous or expressed proteins without need for secondary treatment such as sonication or freeze/thaw
- Formulated for utilization in western blotting, immunoprecipitation and kinase/phosphatase assays
- Reagent is compatible with protease inhibitors, kinase inhibitors and phosphatase inhibitors
Applications:
Western blotting, immunoprecipitation and kinase/phosphatase assays
Specifications
| 50mL | |
| Protein Extraction Reagent | |
| Western blotting, immunoprecipitation and kinase/phosphatase assays | |
| 15° to 30°C |