missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ ColiRollers™ Plating Beads

Used for plating bacteria for transformation molecular cloning applications.

$54.30 - $169.00

Specifications

Content And Storage 15°C to 30°C
Form Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
710133
View Documents Promotion Details
MilliporeSigma™
710133
1 Pack
Each for $54.30
Only null left
 
710134
View Documents Promotion Details
MilliporeSigma™
710134
5/Pk.
Each for $169.00
Only null left
 
Description

Description

EMD Millipore Novagen™ ColiRollers™ Plating Beads provides even distribution of colonies.

  • Specially treated glass beads
  • Eliminates inconsistencies of conventional methods
  • Provides even distribution of colonies
  • Sterile
  • Each package contains beads for 60 to 80 plates
Specifications

Specifications

15°C to 30°C
Solid
Videos
SDS
Documents

Documents