missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ BugBuster™ Master Mix

Catalog No. 714563
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
714563 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 714563 Supplier MilliporeSigma™ Supplier No. 714563

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Allows for recovery of active soluble protein from bacteria.

  • Combines BugBuster protein extraction reagent with Benzonase™ Nuclease and rLysozyme™ Solution in one convenient reagent

Specifications

Formulation BugBuster Protein Extraction Reagent with Benzonase™ Nuclease and rLysozyme™ Solution in one reagent
Quantity 100 mL
Sufficient For 20 g cell paste
For Use With (Application) Recovery of active soluble protein from both Gram-negative and Gram-positive bacteria
Content And Storage Store at 2° to 8°C.