missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ Benzonase™ Nuclease
Description
- Genetically engineered endonuclease from Serratia marcescens
- Degrades all forms of DNA and RNA (single stranded, double stranded, linear and circular) while having no proteolytic activity
- Effective over wide range of conditions and possesses high specific activity
- Rapidly hydrolyzes nucleic acids to makes enzyme for viscosity reduction to reduce processing time and increase yields of protein
- Compatible with BugBuster™ and PopCulture™ Protein Extraction Reagents
- Can be added along with these reagents to eliminate viscosity and remove nucleic acids from E. coli extracts
- Functional between pH 6 and 10 and from 0 to 42°C and requires 1to 2mM Mg2+ for activation
- Active in presence of ionic and non-ionic detergents, reducing agents, PMSF (1mM), EDTA (1mM) and urea
- Activity is inhibited by >150mM monovalent cations, >100mM phosphate, >100mM ammonium sulfate, or >100mM guanidine HCl
- Packaging: Plastic bottle
Compliance
Compliant with FDA
Specifications
Specifications
| Concentration | 25 to 29U/μL |
| For Use With (Application) | Nucleic Acids Removal |
| Formulation | Supplied in 50% glycerol |
| pH Range | 6 to 10 |
| Quantity | 10 kU |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Product Type | Nuclease |
| Purity or Quality Grade | >90% |