Learn More
MilliporeSigma™ Novagen™ BCA Protein Assay Kit
Catalog No. 712853
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
712853 1 Kit
1 options
Catalog No. 712853 Supplier MilliporeSigma™ Supplier No. 712853
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Determines protein concentration in standard assay or microassay configuration.
EMD Millipore Novagen™ BCA Protein Assay Kit is based on biuret reaction, which is the reduction of Cu2+ to Cu+ by proteins in alkaline solution.
  • Sufficient to complete 500 standard-size reactions or or 2500 microscale reactions
  • Used to determine protein concentrationin the range of 20-2000 μg/mL
  • Robust, can be performed in presence of many compounds
  • BSA standard (2 mg/mL) is provided for convenient and reliable preparation of standard curves
Specifications
Sufficient For 500 standard-size reactions (50 μL protein sample plus 1 mL reagent) or 2500 microscale reactions (25 μL protein sample plus 200 μL reagent)
Kit Contents 500 mL BCA Solution, 15 mL 4% Cupric Sulfate, 3 x 1 mL BSA Standard, 2 mg/mL, Poly(dI-dC) solution, Sonicated salmon sperm DNA
Quantitation Range 20 to 2000 μg/mL
Quantity 1 Kit
Storage Requirements Store at room temperature