missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ BacMagic™ 3 Transfection Kit

Catalog No. 723513
Change view
Click to view available options
Quantity:
3 Transfection Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
723513 3 Transfection Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 723513 Supplier MilliporeSigma™ Supplier No. 723513

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Generates recombinant baculoviruses for protein expression in insect cells.

  • Eliminates tedious, time-consuming steps of plaque purification
  • Improves production of secreted and membrane target proteins
  • Improved quality and yield for most target proteins
  • Packaging: Glass Bottle

Specifications

Content And Storage Multiple Storage Conditions
Includes BacMagic-3 DNA, Insect GeneJuice Transfection Reagent, Transfection Control Plasmid
Quantity 3 Transfection Kit
Form Liquid