missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ 293-Free™,Core Transfection Reagent

Catalog No. 721815
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
721815 10 mL
721813 1 mL
721814 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 721815 Supplier MilliporeSigma™ Supplier No. 721815

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for transfection of HEK293 cells grown in suspension culture

  • Polycationic liposomal formulation
  • Ideal for mammalian protein production
  • Derived from non-animal sources
  • Minimal cellular toxicity
  • Sterile ready to use solution
  • Optimized for transient transfections of HEK 293 suspension cultures
  • Compatible with both serum-containing serum-free media
  • Eliminates need for media changes
  • Packaging: Glass Bottle

Specifications

Content And Storage 2°C to -8°C
Quantity 10 mL
Form Liquid