missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Protein Analysis Reagents
- Crosslinking, Labeling and Protein Modification
- Protein Labeling Reagents
- Invitrogen™ NovaFluor™ Antibody Conjugation Kits
Invitrogen™ NovaFluor™ Antibody Conjugation Kits
Catalog No. p-9405003
Change view
Click to view available options
Label or Dye:
NovaFluor™ Blue 510
NovaFluor™ Blue 530
NovaFluor™ Blue 555
NovaFluor™ Blue 585
NovaFluor™ Blue 610/30S
NovaFluor™ Blue 610/70S
NovaFluor™ Blue 660/120S
NovaFluor™ Blue 660/40S
NovaFluor™ Red 660
NovaFluor™ Red 685
NovaFluor™ Red 700
NovaFluor™ Red 710
NovaFluor™ Yellow 570
NovaFluor™ Yellow 590
NovaFluor™ Yellow 610
NovaFluor™ Yellow 660
NovaFluor™ Yellow 690
NovaFluor™ Yellow 700
NovaFluor™ Yellow 730
Product Type:
Conjugation Kit
Emission:
510 nm
530 nm
555 nm
570 nm
585 nm
590 nm
610 nm
660 nm
685 nm
690 nm
700 nm
710 nm
730 nm
Quantity:
100 μg
19 product options available for selection
| Catalog No. | Label or Dye | Product Type | Emission | Quantity |
|---|---|---|---|---|
| K06T04L008 | NovaFluor™ Blue 510 | Conjugation Kit | 510 nm | 100 μg |
| K06T04L002 | NovaFluor™ Blue 530 | Conjugation Kit | 530 nm | 100 μg |
| K06T04L009 | NovaFluor™ Blue 555 | Conjugation Kit | 555 nm | 100 μg |
| K06T04L010 | NovaFluor™ Blue 585 | Conjugation Kit | 585 nm | 100 μg |
| K06T04L003 | NovaFluor™ Blue 610/30S | Conjugation Kit | 610 nm | 100 μg |
| K06T04L011 | NovaFluor™ Blue 610/70S | Conjugation Kit | 610 nm | 100 μg |
| K06T04L004 | NovaFluor™ Blue 660/40S | Conjugation Kit | 660 nm | 100 μg |
| K06T04L012 | NovaFluor™ Blue 660/120S | Conjugation Kit | 660 nm | 100 μg |
| K06T04L017 | NovaFluor™ Red 660 | Conjugation Kit | 660 nm | 100 μg |
| K06T04L018 | NovaFluor™ Red 685 | Conjugation Kit | 685 nm | 100 μg |
| K06T04L019 | NovaFluor™ Red 700 | Conjugation Kit | 700 nm | 100 μg |
| K06T04L020 | NovaFluor™ Red 710 | Conjugation Kit | 710 nm | 100 μg |
| K06T04L005 | NovaFluor™ Yellow 570 | Conjugation Kit | 570 nm | 100 μg |
| K06T04L013 | NovaFluor™ Yellow 590 | Conjugation Kit | 590 nm | 100 μg |
| K06T04L006 | NovaFluor™ Yellow 610 | Conjugation Kit | 610 nm | 100 μg |
| K06T04L007 | NovaFluor™ Yellow 660 | Conjugation Kit | 660 nm | 100 μg |
| K06T04L014 | NovaFluor™ Yellow 690 | Conjugation Kit | 690 nm | 100 μg |
| K06T04L015 | NovaFluor™ Yellow 700 | Conjugation Kit | 700 nm | 100 μg |
| K06T04L016 | NovaFluor™ Yellow 730 | Conjugation Kit | 730 nm | 100 μg |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
Description
NovaFluor Conjugation Kits provide a simple workflow to conjugate a blue, yellow-green or red excitable fluorophore to your antibody of choice in just 3 easy steps. The fluorescence-by-design Phiton platform enables the addition of parameters to your current panels in discovery research and resolves stained populations with near-zero cross excitation, with a lack of emission in the violet channels that plague other fluorophores.
NovaFluor Conjugation Kit provides features including:
- A clean and bright antibody label to increase the number of parameters on conventional and spectral flow cytometers
- Resolve populations with limited/near-zero cross excitation and the lowest spread performance of any blue, yellow-green or red excited fluorescent labels
- Simple 3-step protocol, with 60 sec. of hands-on time for conjugating 10–100 μg of antibody per label
- Up to 70% less excitation when compared to PE tandems, resulting in decreased spillover and higher resolution
3-step conjugation method
- Attach NovaFluor Linker to antibody of choice
- Remove excess NovaFluor Linker from DNA-conjugate antibody
- Anneal NovaFluor to oligo-conjugated antibody
Specifications
Specifications
| For Use With (Application) | Flow Cytometry |
| Quantity | 100 μg |
| Product Type | Conjugation Kit |
| Label or Dye | NovaFluor™ Blue 510 |
| Product Line | NovaFluor |
| Emission | 510 nm |
For Research Use Only. Not for use in diagnostic procedures.