missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ NovaClean™ Lab and Glass Cleaner

Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.
$116.16
Specifications
| Product Type | Filtered Low Ionic Surfactant (0.1 μ) |
|---|---|
| Color-coded | No |
| pH | 7.1 |
| Product Line | NovaClean |
| Scent | Unscented |
Description
Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.
Lab and glass cleaner is a low-ionic detergent, designed for use on cleanroom work-stations, laminar flow benches, acrylic and painted surfaces.
- Ready to use, cleanroom surfactant
- Filtered to 0.1 microns
- Low mobile ions
- Low non-volatile residue levels
- Can be applied directly to a range of surfaces
- Works well with glass, stainless steel, epoxy, acrylic, and vinyl curtains
- Ideal for conductive flooring
- Available in a 1 quart spray bottle and a 1 gallon refill bottle
- Biodegradable
- Free-rinsing
Specifications
| Filtered Low Ionic Surfactant (0.1 μ) | |
| 7.1 | |
| Unscented | |
| Clear | |
| 99°C (210.2°F) | |
| ISO 9001 Certified Facility; Certificate of Analysis | |
| Store at ambient temperature. Avoid direct sunlight. Store in original container and keep tightly closed and seaqled until ready for use. | |
| Reusable | |
| No | |
| Mops and Buckets or Wipes and Sponge | |
| The product is chemically stable. Under normal conditions of storage and use, hazardous reactions will not occur. |
| No | |
| NovaClean | |
| Non-sterile | |
| Not Autoclavable | |
| Avoid breathing vapor or mist and provide adequete ventilation when using. | |
| Clear | |
| 1.025 g/mL at 25°C | |
| Filtered | |
| Ready to use filtered cleanroom detergent, no mobile ions, free-rinsing for cleaning glass, stainless steel, acrylic and painted surfaces. | |
| 0°C (32°F) | |
| 2.3 kPa (17.1 mmHg) |
Safety and Handling
ShelfLife : 24 Months