Learn More
Invitrogen™ Human NOV ELISA Kit
ELISA
$884.00
Specifications
| Accession Number | P48745 |
|---|---|
| Assay Range | 0.041 to 30 ng/mL |
| Assay Sensitivity | 2 pg/mL |
| Calibration Range | 2 pg/mL to 30 ng/mL |
| Assay | Solid-phase sandwich ELISA |
Description
The Human NOV ELISA quantitates Hu NOV in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu NOV. Principle of the method The Human NOV solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
NOV is a member of the CCN family of secreted, cysteine-rich regulatory proteins. The full-length NOV protein contains four structural domains that confer distinct, and sometimes opposing, biological activities. Elevated expression of NOV is associated with certain tumors, including Wilm's tumor and most nephroblastomas. However, in other tumor types and certain cancer cell lines, increased tumorigenicity and proliferation is correlated with decreased NOV expression. Additionally, NOV induces cell adhesion and cell migration by signaling through specific cell-surface integrins, and by binding to heparin sulfate proteoglycans and to fibulin 1C. NOV has also been reported to exert proangiogenic activities.Specifications
| P48745 | |
| 2 pg/mL | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| Human | |
| Colorimetric Microplate Reader | |
| CCN3,IBP-9,IGFBP-9,IGFBP9h,NOV | |
| <10 | |
| HRP | |
| Gene Regulation | |
| 2°C to 8°C | |
| NOV | |
| 4 hrs 45 mins |
| 0.041 to 30 ng/mL | |
| 2 pg/mL to 30 ng/mL | |
| HRP | |
| Other Proteins | |
| This ELISA antibody pair detects human NOV. Other species not determined yet. | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 4856 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| CCN3, IBP-9, IGFBP-9, IGFBP9h, NOV | |
| 1 hrs 20 mins |