missing translation for 'onlineSavingsMsg'
Learn More

Nortropinone hydrochloride, 97%

CAS: 25602-68-0 | C7H12ClNO | 161.629 g/mol

Supplier:  Thermo Scientific Chemicals H6448514

 View more versions of this product

Catalog No. AAH6448514


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nortropinone hydrochloride
194°C to 196°C
MFCD03613582
3693524
nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride
C1CC2CC(=O)CC1N2.Cl
161.629
161.63
Solid
25602-68-0
C7H12ClNO
25 g
Hygroscopic
MZQWQFWRSDNBPV-UHFFFAOYSA-N
8-azabicyclo[3.2.1]octan-3-one;hydrochloride
13091218
≥98%

Chemical Identifiers

25602-68-0
161.629
MZQWQFWRSDNBPV-UHFFFAOYSA-N
13091218
C1CC2CC(=O)CC1N2.Cl
C7H12ClNO
MFCD03613582
nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride
8-azabicyclo[3.2.1]octan-3-one;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only