missing translation for 'onlineSavingsMsg'
Learn More

Nortropinone hydrochloride, 97%

Catalog No. p-7055811
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6448506 5 g
AAH6448514 25 g
AAH6448522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6448506 Supplier Thermo Scientific Chemicals Supplier No. H6448506
Only null left

CAS: 25602-68-0 | C7H12ClNO | 161.629 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25602-68-0
Molecular Formula C7H12ClNO
Molecular Weight (g/mol) 161.629
MDL Number MFCD03613582
InChI Key MZQWQFWRSDNBPV-UHFFFAOYSA-N
Synonym nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride
PubChem CID 13091218
IUPAC Name 8-azabicyclo[3.2.1]octan-3-one;hydrochloride
SMILES C1CC2CC(=O)CC1N2.Cl

Specifications

Melting Point 194°C to 196°C
Quantity 5 g
Beilstein 3693524
Sensitivity Hygroscopic
Formula Weight 161.63
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Nortropinone hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only