missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ North™ Low-Profile Cartridges and Cartridge/P100 Filter Combinations
Description
- All North air-purifying cartridges are approved with all North air-purifying half masks and full-facepiece respirators
- Air-purifying respirators with cartridges provide respiratory protection against gases and vapors by using sorbents to purify the inhaled air
- Cartridges without filters do not remove dust, fibers, and other particles
Certifications
All North cartridges, filters, and cartridge/filter combinations are NIOSH certified to 42CFR84 standards when used with North respirator facepieces.
Compliance
NIOSH approved.
Specifications
Specifications
| Cartridge Type | Filter Cartridge |
| Certifications/Compliance | NIOSH, 42CFR84 |
| Color | Olive/Magenta |
| Product Type | Defender Multipurpose/P100 Combo |
| For Use With (Application) | Industrial applications |
| For Use With (Equipment) | Respirator |
| Quantity | 12/Cs. |