missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Normal Rat Serum, eBioscience™
Description
Description: This normal rat serum can be used to inhibit non-specific binding of rat monoclonal antibodies during staining for flow cytometric analysis or immunohistochemistry. Applications Reported: Normal Rat Serum has been reported for use in intracellular staining followed by flow cytometric analysis, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.
Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.
Specifications
Specifications
| Content And Storage | 4°C |
| Form | Liquid |
| Product Type | Rat Serum |
| Quantity | 100 μL |
| Concentration | Conc. Not Determined |
| Formulation | Whole serum with no preservative |