missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Rat Serum, eBioscience™

Catalog No. 501128948
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501128948 500 μL
501128600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501128948 Supplier Invitrogen™ Supplier No. 24555594
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Control

Description: This normal rat serum can be used to inhibit non-specific binding of rat monoclonal antibodies during staining for flow cytometric analysis or immunohistochemistry. Applications Reported: Normal Rat Serum has been reported for use in intracellular staining followed by flow cytometric analysis, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.

Specifications

Content And Storage 4°C
Form Liquid
Product Type Rat Serum
Quantity 500 μL
Concentration Conc. Not Determined
Formulation Whole serum with no preservative