missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Mouse Serum, eBioscience™

Catalog No. 5015426
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5015426 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5015426 Supplier Invitrogen™ Supplier No. 24554494
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Control

Description: This normal mouse serum can be used to inhibit non-specific binding of mouse monoclonal antibodies during staining for flow cytometric analysis. This product can be used with Product # Product # 14-4777-82, Product # 14-4774-82, and Product # 14-797982. It can also be used in other applications which require normal mouse serum.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.

Specifications

Content And Storage 4°C
Form Liquid
Product Type Normal Mouse Serum
Quantity 500 μL
Concentration Conc. Not Determined
Formulation Whole serum with no preservative
For Use With (Application) Flow Cytometry, Control
Isotype Ig