missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Mouse Serum

Catalog No. 016501
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
016501 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 016501 Supplier Invitrogen™ Supplier No. 016501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Control

This serum is collected from NIH Swiss mouse.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.

Specifications

Content And Storage 4°C
Form Lyophilized
Product Type Normal Mouse Serum
Quantity 5 mL
Concentration Conc. Not Determined
Formulation Whole serum with 0.1% sodium azide
For Use With (Application) Control
Isotype Ig