missing translation for 'onlineSavingsMsg'
Learn More

Normal Goat Serum (10%)

Catalog No. 50062Z
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50062Z 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50062Z Supplier Thermo Fisher Scientific Supplier No. 50062Z*

Normal Goat Serum (10%)

This 10% Normal Goat Serum blocking solution is ready-to-use, in phosphate buffered saline (pH 7.4) with 0.1% sodium azide. This normal goat serum is intended for use as a blocking solution when a second antibody of goat origin is applied in an immunohistochemical staining procedure.

To block non-specific binding from the goat second antibody, incubate the tissue section with 10% Normal Goat Serum for 10 minutes at room temperature prior to the primary antibody incubation step.

Specifications

Content And Storage Store at 2°C to 8°C. Do not freeze.
Species Goat/Caprine
Quantity 100 mL
Shipping Condition Wet Ice
Why is Normal Goat Serum (10%) (Cat. No. 50062Z) blue in color?

The blue color of Normal Goat Serum (10%) (Cat. No. 50062Z) is due to a non-toxic food color. The composition of the product is 10% normal goat serum solution in phosphate-buffered saline, pH 7.4, and 0.1% sodium azide (with blue food coloring added).

I used a neuron-specific antibody to label my neurons. How can I reduce non-specific antibody binding?

A blocking step should be performed to reduce fluorescence due to non-specific antibody binding. A common blocking step is the addition of a 2-5% solution of bovine serum albumin (fraction V defatted BSA). Another approach employs the addition of a 5-10% solution of serum from the species in which the secondary antibodies were raised. For example, when using goat anti-mouse IgG secondary antibodies, samples may be effectively blocked with 5-10% normal goat serum. To further reduce background fluorescence, the Image-iT FX Signal Enhancer can be included as a pre-blocking step to decrease non-specific labeling due to charge interactions between the dyes on the conjugates and the cellular constituents.
If you are using a secondary antibody make sure that the species of the antibody is not the same as the species of the sample. For example do not use an anti-mouse secondary antibody on mouse tissue.
Titrate the antibody to the lowest concentration you can use and still get adequate signal.
Try using a fluorescently tagged primary antibody because it should give reduced background but be aware this can reduce signal intensity.

After labeling with my antibody, I am seeing non-specific background binding in my cells or tissue. What could be the cause?

There can be many causes, including insufficient blocking, too high a concentration of the primary or secondary antibody, or degraded primary or secondary antibody. A “no-primary antibody” control can help determine if the secondary antibody is at fault. Otherwise, we recommend trying more stringent blocking or lower concentrations of primary and secondary antibodies.

What concentration of my antibody should I use for cell analysis?

An optimal concentration may be between 1-10 µg/mL for cell and tissue labeling for microscopy, or 0.2-5 µg/mL for flow cytometry. A range of concentrations should be tested to determine what is optimal.

I know I need to block my samples against non-specific protein binding of my antibodies, but which blocker should I use?

For blocking cells or tissues you may use 2-5% bovine serum albumin (fraction V, defatted BSA) or 5-10% normal serum of the species matching the host species of the secondary antibody. Other options include a mixture of BSA and serum or other purified proteins. We offer a ready-made blocking solution, BlockAid Blocking Solution (Cat. No. B10710), which is not species-specific. For cell or tissue samples, avoid the use of non-fat dry milk as a blocking agent as it contains a high level of phosphoproteins, histones, and biotin.

For Laboratory Use