missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norlaudanosine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America N091825G
Specifications
| Norlaudanosine Hydrochloride | |
| 218°C | |
| C20H26ClNO4 | |
| 25 g | |
| VMPLLPIDRGXFTQ-UHFFFAOYNA-N | |
| 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride | |
| 16667431 | |
| ≥98.0% (HPLC,N) |
| 6429-04-5 | |
| White | |
| MFCD00035267 | |
| Tetrahydropapaverine Hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride | |
| COC1=C(C=C(C=C1)CC2C3=CC(=C(C=C3CCN2)OC)OC)OC.Cl | |
| 379.88 | |
| 379.88 | |
| Crystalline Powder |
Chemical Identifiers
| 6429-04-5 | |
| 379.88 | |
| VMPLLPIDRGXFTQ-UHFFFAOYNA-N | |
| 16667431 | |
| COC1=C(C=C(C=C1)CC2C3=CC(=C(C=C3CCN2)OC)OC)OC.Cl |
| C20H26ClNO4 | |
| MFCD00035267 | |
| Tetrahydropapaverine Hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride | |
| 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride |
Safety and Handling
RTECSNumber : NX5070700
TSCA : No