accessibility menu, dialog, popup

UserName

CAS RN 6429-04-5

IUPAC Name: 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Synonyms: 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Molecular Weight (g/mol): 379.88
Molecular Formula: C20H26ClNO4
InChi Key: VMPLLPIDRGXFTQ-UHFFFAOYNA-N
SMILES: [Cl-].COC1=CC=C(CC2[NH2+]CCC3=CC(OC)=C(OC)C=C23)C=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results