missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015291
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015291 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015291 Supplier Merck Emd Millipore Supplier No. PHR16141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Norfloxacin is a synthetic broad-spectrum antibacterial drug that is effective against various gram positive and gram negative bacteria. Its major application is in the treatment of urinary tract infections.

Specifications

CAS 70458-96-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C16H18 FN3O3
MDL Number MFCD00079532
Synonym Norfloxacin; 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid; 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-piperazino-3-quinolinecarboxylic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 319.33
Grade Certified Reference Material