missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin, 98% (TLC), MilliporeSigma™ Supelco™

Catalog No. p-7187492
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015293 1 g
111015294 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015293 Supplier Merck Emd Millipore Supplier No. N98901G
Only null left

Norfloxacin is a broad spectrum antibiotic, which is used against a wide variety of gram positive and gram negative bacteria.

Featured Industries: Environmental Pharmaceutical (small molecule)

Specifications

CAS 70458-96-7
Physical Form Neat
Linear Formula C16H18 FN3O3
MDL Number MFCD00079532
Synonym 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid; 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-piperazino-3-quinolinecarboxylic acid
Type Analytical Standard
Quantity 1 g
Formula Weight 319.33
Percent Purity ≥98% (TLC)
Grade Analytical Standard