missing translation for 'onlineSavingsMsg'
Learn More

Norethisterone, Thermo Scientific Chemicals

Catalog No. AC262890010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC262890010 1 g
AC262890050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC262890010 Supplier Thermo Scientific Chemicals Supplier No. 262890010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68-22-4
Molecular Formula C20H26O2
Molecular Weight (g/mol) 298.42
InChI Key VIKNJXKGJWUCNN-XGXHKTLJSA-N
Synonym norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
PubChem CID 6230
ChEBI CHEBI:7627
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34

Specifications

Melting Point 205.0°C to 206.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 3 h)
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, dioxane
Formula Weight 298.42
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Norethisterone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Suspected of causing cancer. May damage fertility. May damage the unborn child. May cause harm to breast-fed children.
Precautionary Statement GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention. Avoid contact during pregnancy/while nursing.
EINECSNumber 200-681-6

RUO – Research Use Only