missing translation for 'onlineSavingsMsg'
Learn More

Norethisterone Acetate 98.0+%, TCI America™

Supplier:  TCI America N04501G

 View more versions of this product

Catalog No. N04501G


Only null left

Description

Description

Specifications

Specifications

Norethisterone Acetate
White
MFCD04039850
norethisterone acetate, 19-norethindrone acetate, norethindrone acetate, enta, orlutate, norlutin acetate, norlutate, 19-norethisterone acetate, primolut-nor, aygestin
CC(=O)OC1(CCC2C1(CCC3C2CCC4=CC(=O)CCC34)C)C#C
340.463
CHEBI:7628
≥98.0% (HPLC,T)
51-98-9
C22H28O3
1 g
IMONTRJLAWHYGT-ZCPXKWAGSA-N
[(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
5832
340.46
Crystalline Powder

Chemical Identifiers

51-98-9
340.463
IMONTRJLAWHYGT-ZCPXKWAGSA-N
5832
[(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
C22H28O3
MFCD04039850
norethisterone acetate, 19-norethindrone acetate, norethindrone acetate, enta, orlutate, norlutin acetate, norlutate, 19-norethisterone acetate, primolut-nor, aygestin
CHEBI:7628
CC(=O)OC1(CCC2C1(CCC3C2CCC4=CC(=O)CCC34)C)C#C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : RC8965000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions