accessibility menu, dialog, popup

UserName

CAS RN 51-98-9

IUPAC Name: (1R,3aS,3bR,9aR,9bS,11aS)-1-ethynyl-11a-methyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Synonyms: ENTA norlutin A miniphase progylut norethysterone acetate milli-anovlar monogest gestakadin norethisteron acetate primolut-nor
Molecular Weight (g/mol): 340.46
Molecular Formula: C22H28O3
InChi Key: IMONTRJLAWHYGT-ZCPXKWAGSA-N
SMILES: CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)