missing translation for 'onlineSavingsMsg'
Learn More

Norethisterone Acetate 98.0+%, TCI America™

Catalog No. N0450100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0450100MG 100 mg
N04501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0450100MG Supplier TCI America Supplier No. N0450100MG

Chemical Identifiers

CAS 51-98-9
Molecular Formula C22H28O3
Molecular Weight (g/mol) 340.463
MDL Number MFCD04039850
InChI Key IMONTRJLAWHYGT-ZCPXKWAGSA-N
Synonym norethisterone acetate, 19-norethindrone acetate, norethindrone acetate, enta, orlutate, norlutin acetate, norlutate, 19-norethisterone acetate, primolut-nor, aygestin
PubChem CID 5832
ChEBI CHEBI:7628
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)OC1(CCC2C1(CCC3C2CCC4=CC(=O)CCC34)C)C#C

Specifications

Color White
Quantity 100 mg
Formula Weight 340.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Norethisterone Acetate
RTECSNumber RC8965000
TSCA No