missing translation for 'onlineSavingsMsg'
Learn More

Norethindrone, MP Biomedicals™

Catalog No. p-4405164
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15594683 0.25 g
ICN15594691 1 g
ICN15594694 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15594683 Supplier MP Biomedicals Inc Supplier No. 0215594683
Only null left

Chemical Identifiers

CAS 68-22-4
Molecular Formula C20H26O2
Molecular Weight (g/mol) 298.426
InChI Key VIKNJXKGJWUCNN-XGXHKTLJSA-N
Synonym norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
PubChem CID 6230
ChEBI CHEBI:7627
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34

Specifications

Color White
Quantity 0.25 g
Formula Weight 298.4
Physical Form Powder
Chemical Name or Material Norethindrone