missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nordoxepin hydrochloride, 98% mixed isomers basis (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 2887-91-4 |
| Physical Form | Powder |
| Linear Formula | C18H19 NO · HCl |
| MDL Number | MFCD00135797 |
| Synonym | 11(6 H)-(3-[Methylamino]propylidene)dibenz[b,e]oxepine |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 301.81 |
| Percent Purity | ≥98% mixed isomers basis (GC) |
| Grade | Analytical Standard |