missing translation for 'onlineSavingsMsg'
Learn More

Norbornane 98.0+%, TCI America™

Catalog No. N051710G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N051710G 10 g
N051725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N051710G Supplier TCI America Supplier No. N051710G

Chemical Identifiers

CAS 279-23-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00074822
InChI Key UMRZSTCPUPJPOJ-UHFFFAOYSA-N
Synonym Bicyclo[2.2.1]heptane
PubChem CID 9233
IUPAC Name bicyclo[2.2.1]heptane
SMILES C1CC2CCC1C2

Specifications

Melting Point 85°C
Color White
Boiling Point 106°C
Quantity 10 g
Formula Weight 96.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Norbornane
RTECSNumber RB6945000
TSCA No