missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Noradrenaline Competitive ELISA Kit

Catalog No. EEL010
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL010 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL010 Supplier Invitrogen™ Supplier No. EEL010

ELISA

The multispecies Noradrenaline/Norepinephrine (NA/NE) ELISA quantitates NA/NE in serum, plasma, and other biological fluids Principle of the method: The Competitive multispecies ELISA research-use-only kit is designed to quantitatively measure Noradrenaline/Norepinephrine (NA/NE) independent of species. A NA/NE standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to NA/NE is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme-substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of NA/NE in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Norepinephrine (NE), also called noradrenalin (NA), is an organic chemical in the catecholamine family that functions in the brain and body as both a hormone and neurotransmitter. NA/NE acts on target cells by binding and activating adrenergic receptors located on the cell surface to mobilize the brain and body for action. Its differs structurally from epinephrine in that the methyl group attached to its nitrogen is replaced by a hydrogen atom in NE.

Specifications

Assay Range 0.31-20 ng/mL
Assay Sensitivity 0.19 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Catecholamine
Sample Type Plasma, Serum
Target Species All Species
For Use With (Equipment) Absorbance Microplate Reader
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Noradrenaline
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less