missing translation for 'onlineSavingsMsg'
Learn More

NOP132 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7141691
Encompass
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501563308 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501563308 Supplier Bethyl Laboratories Inc Supplier No. A302722AT
Only null left

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Plays an essential role in the survival of diffuse-type gastric cancer cells. Acts as a nucleolar anchoring protein for DDX47. May be involved in regulation of gene expression at the post-transcriptional level or in ribosome biogenesis in cancer cells. {ECO:0000250}.

Specifications

Antigen NOP132
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene NOL8
Gene Accession No. Q76FK4
Gene Alias nucleolar protein 8, bA62C3.4, bA62C3.3, C9orf34, NOP132
Gene Symbols NOL8
Host Species Rabbit
Immunogen Between 200 and 250
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55035
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less