missing translation for 'onlineSavingsMsg'
Learn More

Nonyl Chloroformate 95.0+%, TCI America™

Catalog No. B026225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B026225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B026225ML Supplier TCI America Supplier No. B026225ML
Only null left

Chemical Identifiers

CAS 57045-82-6
Molecular Formula C10H19ClO2
Molecular Weight (g/mol) 206.71
MDL Number MFCD00059485
InChI Key VKOFPDUSOTWTPX-UHFFFAOYSA-N
Synonym Chloroformic Acid Nonyl Ester
PubChem CID 544663
IUPAC Name nonyl carbonochloridate
SMILES CCCCCCCCCOC(=O)Cl

Specifications

Color Colorless
Quantity 25 mL
UN Number 3277
Formula Weight 206.71
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Nonyl Chloroformate