accessibility menu, dialog, popup

UserName

CAS RN 57045-82-6

H3COOCl

CAS RN 57045-82-6

IUPAC Name: nonyl carbonochloridate
Synonyms: nonyl carbonochloridate
Molecular Weight (g/mol): 206.71
Molecular Formula: C10H19ClO2
InChi Key: VKOFPDUSOTWTPX-UHFFFAOYSA-N
SMILES: CCCCCCCCCOC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results