missing translation for 'onlineSavingsMsg'
Learn More

Nonyl beta-D-Glucopyranoside 98.0+%, TCI America™

Catalog No. N09091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09091G Supplier TCI America Supplier No. N09091G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69984-73-2
Molecular Formula C15H30O6
Molecular Weight (g/mol) 306.399
MDL Number MFCD00063300
InChI Key QFAPUKLCALRPLH-UXXRCYHCSA-N
Synonym nonyl beta-d-glucopyranoside, b-nonylglucoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy tetrahydro-2h-pyran-3,4,5-triol, b-d-glucopyranoside, nonyl, nonyl-beta-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy oxane-3,4,5-triol, n-nonyl-beta-d-glucopyranoside, bng, beta-d-glucopyranoside, nonyl, 4gby
PubChem CID 155448
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol
SMILES CCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 70°C
Color White
Quantity 1 g
Formula Weight 306.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Nonyl beta-D-Glucopyranoside
EINECSNumber (5)-3641
TSCA No
Recommended Storage Freezer