missing translation for 'onlineSavingsMsg'
Learn More

Nonyl Acetate 99.0+%, TCI America™

Catalog No. A0041500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0041500ML 500 mL
A004125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0041500ML Supplier TCI America Supplier No. A0041500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 143-13-5
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00027340
InChI Key GJQIMXVRFNLMTB-UHFFFAOYSA-N
Synonym acetic acid, nonyl ester, n-nonyl acetate, n-nonanyl acetate, acetic acid n-nonyl ester, nonanol acetate, pelargonyl acetate, n-nonyl ethanoate, acetate c-9, nonyl ethanoate, 1-acetoxynonane
PubChem CID 8918
ChEBI CHEBI:87511
IUPAC Name nonyl acetate
SMILES CCCCCCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 228°C
Quantity 500 mL
Formula Weight 186.30
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonyl Acetate
EINECSNumber (2)-0735
RTECSNumber AJ1382500
TSCA Yes