accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 143-13-5

IUPAC Name: nonyl acetate
Synonyms: nonyl acetate
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: GJQIMXVRFNLMTB-UHFFFAOYSA-N
SMILES: CCCCCCCCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results