missing translation for 'onlineSavingsMsg'
Learn More

Nonaqueous Buffer Stock Solution B, Ricca Chemical

Catalog No. 546332
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
546332 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 546332 Supplier Ricca Chemical Company Supplier No. 546332

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Acid Number and Neutralization Number of Petroleum Products

Specifications

Chemical Name or Material Nonaqueous Buffer Stock Solution B
Name Note For Acid Number and Neutralization Number of Petroleum Products
CAS 67-63-0, 554-84-7, 1310-58-3, 107-15-3
CAS Min % 94.0, 3.33, 0.4, 0.02
Color Amber
CAS Max % 98.0, 3.47, 0.41, 0.02
Concentration 1X
For Use With (Application) Water Analysis
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Amber Glass Bottle
Grade Laboratory
Show More Show Less