missing translation for 'onlineSavingsMsg'
Learn More

Nonanoic acid, 97%

Catalog No. p-7025862
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21568AK 250 mL
AAB21568AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB21568AK Supplier Thermo Scientific Chemicals Supplier No. B21568AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-05-0 | C9H18O2 | 158.241 g/mol

Nonanoic acid is used in the preparation of plasticizers and lacquers. It is commonly used in conjunction with glyphosate, for a quick burn-down effect in the control of weeds in turfgrass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonanoic acid is used in the preparation of plasticizers and lacquers. It is commonly used in conjunction with glyphosate, for a quick burn-down effect in the control of weeds in turfgrass.

Solubility
Soluble in water at 20°C,0.26g/L. Soluble in alcohol and dipropylene glycol.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 112-05-0
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00004433
InChI Key FBUKVWPVBMHYJY-UHFFFAOYSA-N
Synonym pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
PubChem CID 8158
ChEBI CHEBI:29019
IUPAC Name nonanoic acid
SMILES CCCCCCCCC(=O)O

Specifications

Melting Point 10°C to 12°C
Density 0.905
pH 3.5
Boiling Point 253°C to 255°C
Flash Point 140°C (284°F)
Odor Unpleasant
Refractive Index 1.4315
Quantity 250 mL
UN Number UN3265
Beilstein 1752351
Merck Index 14,7070
Solubility Information Soluble in water at 20°C,0.26g/L. Soluble in alcohol and dipropylene glycol.
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material Nonanoic acid
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 203-931-2
RTECSNumber RA6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only