missing translation for 'onlineSavingsMsg'
Learn More

Nonafluorobutyl Iodide 98.0+%, TCI America™

Catalog No. N049925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N049925G 25 g
N0499100G 100 g
N0499500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N049925G Supplier TCI America Supplier No. N049925G

Chemical Identifiers

CAS 423-39-2
Molecular Formula C4F9I
Molecular Weight (g/mol) 345.934
MDL Number MFCD00001062
InChI Key PGRFXXCKHGIFSV-UHFFFAOYSA-N
Synonym perfluorobutyl iodide, nonafluoro-1-iodobutane, nonafluorobutyl iodide, perfluorobutyliodide, n-nonafluorobutyl iodide, perfluoro-n-butyl iodide, 1-iodoperfluorobutane, ccris 9008, 1-iodononafluorobutane, butane, 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodo
PubChem CID 67917
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
SMILES C(C(C(F)(F)I)(F)F)(C(F)(F)F)(F)F

Specifications

Color Red-Yellow
Boiling Point 67°C
Quantity 25 g
Formula Weight 345.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonafluorobutyl Iodide
RTECSNumber EK5360000
TSCA Yes