Learn More
Nonafluorobutanesulfonyl fluoride, 90+%
CAS: 375-72-4 | C4F10O2S | 302.086 g/mol
Supplier: Thermo Scientific Chemicals B2132222
Description
Benzynes were generated from o-(trimethylsilyl) phenols using nonafluorobutanesulfonyl fluoride (NfF) by a domino process.Nonafluorobutanesulfonyl fluoride (nonaflyl fluoride, C 4 F 9 SO 2 F, NfF) is the most widely used commercially available reagent for the synthesis of nonaflates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzynes were generated from o-(trimethylsilyl) phenols using nonafluorobutanesulfonyl fluoride (NfF) by a domino process.Nonafluorobutanesulfonyl fluoride (nonaflyl fluoride, C 4 F 9 SO 2 F, NfF) is the most widely used commercially available reagent for the synthesis of nonaflates.
Solubility
Hydrolyses in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, baese..
Specifications
| Nonafluorobutanesulfonyl fluoride | |
| -110°C | |
| 65°C to 66°C | |
| C4F10O2S | |
| MFCD00007422 | |
| UN3265 | |
| Moisture sensitive | |
| Hydrolyses in water. | |
| C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F | |
| 302.086 | |
| 302.09 |
| 375-72-4 | |
| 1.75 | |
| None | |
| 1.281 | |
| 100 g | |
| 1813589 | |
| nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro | |
| LUYQYZLEHLTPBH-UHFFFAOYSA-N | |
| 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride | |
| 67814 | |
| ≥90% |
Chemical Identifiers
| 375-72-4 | |
| 302.086 | |
| LUYQYZLEHLTPBH-UHFFFAOYSA-N | |
| 67814 | |
| C(C(C(F)(F)S(=O)(=O)F)(F)F)(C(F)(F)F)(F)F |
| C4F10O2S | |
| MFCD00007422 | |
| nonafluorobutanesulfonyl fluoride, perfluorobutanesulfonyl fluoride, perfluoro-1-butanesulfonyl fluoride, perfluorobutanesulfonylfluoride, perfluorobutylsulfonyl fluoride, nonafluorobutanesulphonyl fluoride, perfluoro n-butylsulfonyl fluoride, nonafluorobutane-1-sulfonyl fluoride, nonafluoro-1-butanesulfonyl fluoride, 1-butanesulfonyl fluoride, 1,1,2,2,3,3,4,4,4-nonafluoro | |
| 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H370
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-792-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only