missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonadecane 99.0+%, TCI America™
$96.26 - $687.28
Chemical Identifiers
| CAS | 629-92-5 |
|---|---|
| Molecular Formula | C19H40 |
| Molecular Weight (g/mol) | 268.529 |
| MDL Number | MFCD00009012 |
| InChI Key | LQERIDTXQFOHKA-UHFFFAOYSA-N |
| Synonym | n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl |
| PubChem CID | 12401 |
| ChEBI | CHEBI:32927 |
| IUPAC Name | nonadecane |
| SMILES | CCCCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 629-92-5 | |
| 268.529 | |
| LQERIDTXQFOHKA-UHFFFAOYSA-N | |
| 12401 | |
| nonadecane |
| C19H40 | |
| MFCD00009012 | |
| n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl | |
| CHEBI:32927 | |
| CCCCCCCCCCCCCCCCCCC |
Specifications
| 629-92-5 | |
| 330°C | |
| MFCD00009012 | |
| n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl | |
| CCCCCCCCCCCCCCCCCCC | |
| 268.529 | |
| CHEBI:32927 | |
| ≥99.0% (GC) | |
| Nonadecane |
| White-Yellow | |
| C19H40 | |
| 25 g | |
| LQERIDTXQFOHKA-UHFFFAOYSA-N | |
| nonadecane | |
| 12401 | |
| 268.53 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0010
TSCA : Yes