missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Nonadecane | |
| White-Yellow | |
| C19H40 | |
| 25 g | |
| LQERIDTXQFOHKA-UHFFFAOYSA-N | |
| nonadecane | |
| 12401 | |
| 268.53 | |
| Crystalline Lumps |
| 629-92-5 | |
| 330°C | |
| MFCD00009012 | |
| n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl | |
| CCCCCCCCCCCCCCCCCCC | |
| 268.529 | |
| CHEBI:32927 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 629-92-5 | |
| 268.529 | |
| LQERIDTXQFOHKA-UHFFFAOYSA-N | |
| 12401 | |
| nonadecane |
| C19H40 | |
| MFCD00009012 | |
| n-nonadecane, nonadekan, unii-nmy21d3y5t, nmy21d3y5t, unknown branched fragment of phospholipid, ch3-ch2 17-ch3, unknown phospholipid fragment, nonadecane, analytical standard, upl | |
| CHEBI:32927 | |
| CCCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : (2)-0010
TSCA : Yes