missing translation for 'onlineSavingsMsg'
Learn More

Nonadecane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014956
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014956 5 g
111014955 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014956 Supplier Merck Emd Millipore Supplier No. 741585G
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Ignition Point 446°F
CAS 629-92-5
Vapor Pressure 1 mmHg (133.2°C)
Vapor Density 9.27 (vs. air)
Density 0.786 g/mL (at 25°C)
Flash Point 100°C
Physical Form Neat
Linear Formula CH3(CH2)17 CH3
MDL Number MFCD00009012
Melting Point 30°C to 34°C; 32°C to 34°C
Boiling Point 330°C
Type Analytical Standard
Quantity 5 g
Formula Weight 268.52
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less