missing translation for 'onlineSavingsMsg'
Learn More

Nonadecafluorodecanenitrile 98.0+%, TCI America™

Catalog No. N10951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10951G Supplier TCI America Supplier No. N10951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 379215-40-4
Molecular Formula C10F19N
Molecular Weight (g/mol) 495.087
MDL Number MFCD00045610
InChI Key QXBUHLMUCFBZOL-UHFFFAOYSA-N
Synonym Perfluorodecanonitrile, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanenitrile
PubChem CID 2776067
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanenitrile
SMILES C(#N)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 495.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonadecafluorodecanenitrile