missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N^w,N^w'-Di-Boc-Nalpha-Fmoc-L-arginine, 95%
CAS: 143824-77-5
$154.95 - $1554.67
Chemical Identifiers
| CAS | 143824-77-5 |
|---|---|
| MDL Number | MFCD00237008 |
| Synonym | Fmoc-Arg(Boc)2-OH; (S,E)-5-[1,2-Bis(tert-butoxycarbonyl)guanidino]-2-(Fmoc-amino)pentanoic acid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH63784MD
|
Thermo Scientific Chemicals
H63784MD |
250 mg |
Each for $154.95
|
|
|||||
|
AAH6378403
|
Thermo Scientific Chemicals
H6378403 |
1 g |
Each for $443.95
|
|
|||||
|
AAH6378406
|
Thermo Scientific Chemicals
H6378406 |
5 g |
Each for $1,554.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 143824-77-5 | |
| MFCD00237008 | |
| Fmoc-Arg(Boc)2-OH; (S,E)-5-[1,2-Bis(tert-butoxycarbonyl)guanidino]-2-(Fmoc-amino)pentanoic acid |
Specifications
| 143824-77-5 | |
| Fmoc-Arg(Boc)2-OH; (S,E)-5-[1,2-Bis(tert-butoxycarbonyl)guanidino]-2-(Fmoc-amino)pentanoic acid | |
| 596.67 | |
| Solid |
| MFCD00237008 | |
| 250 mg | |
| 95% | |
| Nomega,Nomega'-Di-Boc-Nalpha-Fmoc-L-arginine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only