missing translation for 'onlineSavingsMsg'
Learn More

N^w,N^w'-Di-Boc-Nalpha-Fmoc-L-arginine, 95%

CAS: 143824-77-5

Supplier:  Thermo Scientific Chemicals H6378403

 View more versions of this product

Catalog No. AAH6378403


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nomega,Nomega'-Di-Boc-Nalpha-Fmoc-L-arginine
MFCD00237008
1 g
95%
143824-77-5
Fmoc-Arg(Boc)2-OH; (S,E)-5-[1,2-Bis(tert-butoxycarbonyl)guanidino]-2-(Fmoc-amino)pentanoic acid
596.67
Solid

Chemical Identifiers

143824-77-5
MFCD00237008
Fmoc-Arg(Boc)2-OH; (S,E)-5-[1,2-Bis(tert-butoxycarbonyl)guanidino]-2-(Fmoc-amino)pentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only