missing translation for 'onlineSavingsMsg'
Learn More

Nomega-Nitro-L-arginine 98.0+%, TCI America™

Catalog No. p-7133917
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06605G 5 g
N066025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06605G Supplier TCI America Supplier No. N06605G
Only null left

Chemical Identifiers

CAS 2149-70-4
Molecular Formula C6H13N5O4
Molecular Weight (g/mol) 219.201
MDL Number MFCD00007033
InChI Key MRAUNPAHJZDYCK-BYPYZUCNSA-N
Synonym h-arg no2-oh, ng-nitro-l-arginine, l-nna, nitroarginine, n-nitro-l-arginine, nomega-nitro-l-arginine, n g-nitro-l-arginine, ng-no2-l-arg, n-omega-nitro-l-arginine, n g-nitroarginine
PubChem CID 440005
ChEBI CHEBI:27960
SMILES C(CC(C(=O)O)N)CN=C(N)N[N+](=O)[O-]

Specifications

Color White
Quantity 5 g
Formula Weight 219.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nomega-Nitro-L-arginine
RTECSNumber RM2982100
TSCA Yes