accessibility menu, dialog, popup

UserName

CAS RN 2149-70-4

IUPAC Name: (2S)-2-azaniumyl-5-{[iminiumyl(nitroamino)methyl]amino}pentanoate
Synonyms: nitroarginine
Molecular Weight (g/mol): 220.21
Molecular Formula: C6H14N5O4
InChi Key: MRAUNPAHJZDYCK-BYPYZUCNSA-O
SMILES: [NH3+][C@@H](CCCNC(=[NH2+])N[N+]([O-])=O)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)