missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554102 Supplier Invitrogen™ Supplier No. PA554102
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: ITDKSLCGSG EKYIPPHVRQ AEETVDFKKK EELERLKKHV KGLLNRLSEP NMASISGQLE ELYMAHSRKD MNDTLTSALM GACVTASAMP SRLMMEHVLL VSILHHTVGI EVGAHFLEAV VRKFDAIYKY GSE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 77%.

Proteins that contain MIF4G (middle of eIF4G (MIM 600495)) and/or MA3 domains, such as NOM1, function in protein translation. These domains include binding sites for members of the EIF4A family of ATP-dependent DEAD box RNA helicases (see EIF4A1; MIM 602641) (Simmons et al., 2005 [PubMed 15715967]).[supplied by OMIM, Mar 2008].

Specifications

Antigen NOM1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOM1
Gene Accession No. Q5C9Z4
Gene Alias C7orf3; D5Kng1; Gm1040; Nom1; Nucleolar MIF4G domain-containing protein 1; nucleolar protein with MIF4G domain 1; PPP1R113; protein phosphatase 1, regulatory subunit 113; SGD1; SGD1 homolog
Gene Symbols NOM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOM1. Recombinant protein control fragment (Product #RP-92761).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64434
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less